missing translation for 'onlineSavingsMsg'
Learn More
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals Product Code.: 10674672

Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals

Product Code. 10674672
1 L, Glass Bottle
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10674672

Brand: Thermo Scientific Acros 176630010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-99-9, 128-37-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108.4°C
Density 0.8800g/mL
Boiling Point 66.0°C
Quantity 1 L
Flash Point −21°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.407
Assay Percent Range 99+%
Residue after Evaporation 0.005% max.
Specific Gravity 0.88
Viscosity 0.55 mPa.s (20°C)
Beilstein 17, 10
Water 0.05% max. (Coulometric)
Fieser 01,1140; 02,398; 03,278; 05,649; 06,570
Merck Index 15, 9356
Solubility Information Solubility in water: miscible. Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride and acetone
Formula Weight 72.11
Percent Purity ≥99%
Grade Extra Pure
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetrahydrofuran
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H225-Highly flammable liquid and vapour.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran, 99+%, extra pure, stabilized with BHT, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.