missing translation for 'onlineSavingsMsg'
Learn More
compliance-icons compliance-icons

Tetrahydrofuran, BAKER ANALYZED HPLC, J.T. Baker™

Product Code. 10461003
2.5 L, Glass
Click to view available options
Quantity:
2.5 L
Packaging:
Glass
This item is not returnable. View return policy
This item is not returnable. View return policy

Get rapid, reproducible performance and separation results. Tetrahydrofuran, BAKER ANALYZED HPLC, J.T. Baker™ is an application-optimized, function-tested HPLC reagent/solvent that maximizes throughput and ease-of-use without sacrificing efficiency or reproducibility.

BAKER ANALYZED HPLC Reagents are high purity solvents, buffer salts, and mobile phase modifiers for use in analytical and preparative separations. They are optimized for research and quality control applications, including high performance liquid chromatography, gel permeation chromatography and UV-spectrophotometric analysis.

  • Recalibration or adjustments due to lot changes are minimized for optimum performance from sensitive instrumentation
  • Low backgrounds free of extraneous peaks
  • Low UV absorbance in critical ranges - assured through UV absorbance testing at a variety of points and through gradient elution testing
  • Fluorescence testing for trace impurities which would cause interference
  • Function-tested for: assay, water, minimal residue after evaporation and UV absorbance and fluorescence in critical ranges
  • Lot to lot consistency, with an actual Lot Analysis on the label
  • Innovative packaging options to assure solvent quality to the point-of-use

Specifications

Melting Point -108.3°C
Absorbance Max. 1.0 at 230nm, Max. 0.15 at 260nm, Max. 0.08 at 270nm, Max. 0.02 at 310nm
CAS 109-99-9
Color Colorless
Density 0.889g/cm3
Boiling Point 65°C
Quantity 2.5 L
Explosion Limit 1.8 - 11.8 Vol %
Flash Point −20°C
Ignition Point 321°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 3 ppm
Molecular Formula C4H8O
Linear Formula OCH2CH2CH2CH2
MDL Number MFCD00005356
Vapor Pressure 21.60 kPa
Water Max. 0.02%
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
Solubility Information Miscible in water
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
SMILES C1CCOC1
IUPAC Name oxolane
Molecular Weight (g/mol) 72.11
PubChem CID 8028
ChEBI CHEBI:26911
Formula Weight 72.11g/mol
Percent Purity ≥99.8%
Grade For use in High Performance Liquid Chromatography
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrahydrofuran, BAKER ANALYZED HPLC, J.T. Baker™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.