missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrofuran, BAKER ANALYZED, J.T. Baker™

Product Code. 10425191
Click to view available options
Quantity:
1 L
5 L
25 L
Packaging:
EcoTainer Metal Safety Can
Glass
Steel
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality grade chemical stabilized with 0.025% BHT for laboratory and specialized industrial use.

  • Label provides an actual Lot Analysis as a guarantee of purity and consistency
  • Analytical methods used for these determinations are based on standard test procedures described in technical publications
  • Improvements developed by J.T. Baker are employed to achieve a higher degree of reliability

Chemical Identifiers

CAS 109-99-9
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00005356
InChI Key WYURNTSHIVDZCO-UHFFFAOYSA-N
Synonym tetrahydrofuran, furan, tetrahydro, butylene oxide, furanidine, hydrofuran, tetramethylene oxide, oxacyclopentane, 1,4-epoxybutane, tetrahydrofuranne, tetraidrofurano
PubChem CID 8028
ChEBI CHEBI:26911
IUPAC Name oxolane
SMILES C1CCOC1

Specifications

Melting Point -108.5°C
Color Colorless
Density 0.880 to 0.883 g/mL at 25°C
Boiling Point 65°C to 67°C
Quantity 1 L
Explosion Limit 1.5 - 12 Vol %
Flash Point −21°C
Ignition Point 230°C
Packaging Glass
Physical Form Liquid
Residue after Evaporation Max. 0.002%
Linear Formula OCH2CH2CH2CH2
Water Max. 0.02%
Solubility Information Fully miscible in water
Formula Weight 72.11g/mol
Percent Purity ≥99%
Grade BAKER ANALYZED™
Chemical Name or Material Tetrahydrofuran
Show More Show Less
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'