missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydrofuran, for HPLC, Unstabilised, Fisher Chemical™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Plastic-coated Glass Bottle
Description
C4H8O, CAS Number-109-99-9, butylene oxide, tetraidrofurano, 1,4-epoxybutane, furanidine, oxacyclopentane, tetrahydrofuranne, hydrofuran, furan, tetrahydro, tetrahydrofuran, tetramethylene oxide, 2.5L, 66 deg.C, CHEBI:26911, Colorless, 72.11g/mol, WYURNTSHIVDZCO-UHFFFAOYSA-N, oxolane, -108.4 deg.C
Specifications
Specifications
pH | 7 to 8 |
Melting Point | -108.4°C |
Color | Colorless |
Boiling Point | 66°C |
Quantity | 1 L |
Packaging | Plastic-coated Glass Bottle |
Physical Form | Liquid |
Vapor Pressure | 200mbar at 20°C |
Viscosity | 0.55cp at 20°C |
Formula Weight | 72.11g/mol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'