missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrahydropyran-4-carboxylic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 87°C to 89°C |
Color | White |
Quantity | 5 g |
Beilstein | 112249 |
Formula Weight | 130.14 |
Percent Purity | 98% |
Physical Form | Solid |
Chemical Name or Material | Tetrahydropyran-4-carboxylic acid |
Safety and Handling
Product Identifier
- Tetrahydropyran-4-carboxylic acid
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : Tetrahydropyran-4-carboxylic acid
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'