missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophen-3-one, 97%, Thermo Scientific Chemicals

Product Code. 11484453
Click to view available options
Quantity:
5 g
10 g
25 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1003-04-9
Molecular Formula C4H6OS
Molecular Weight (g/mol) 102.15
MDL Number MFCD00005412
InChI Key DSXFPRKPFJRPIB-UHFFFAOYSA-N
Synonym tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
PubChem CID 61252
IUPAC Name thiolan-3-one
SMILES O=C1CCSC1

Specifications

Density 1.192
Boiling Point 174°C to 175°C
Flash Point 77°C (170°F)
Odor Characteristic
Refractive Index 1.529
Quantity 250 g
Beilstein 105201
Formula Weight 102.16
Percent Purity 97%
Chemical Name or Material Tetrahydrothiophen-3-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.