missing translation for 'onlineSavingsMsg'
Learn More

Tetrahydrothiophen-3-one, 98%, Thermo Scientific™

Product Code. 11985781
Click to view available options
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 1003-04-9
Density 1.1990g/mL
Boiling Point 174°C to 175°C
Flash Point 78°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Molecular Formula C4H6OS
MDL Number MFCD00005412
Quantity 5g
Specific Gravity 1.199
Synonym tetrahydrothiophen-3-one, 3-thiophanone, dihydrothiophen-3 2h-one, dihydro-3 2h-thiophenone, 3-thiacyclopentanone, 3-oxotetrahydrothiophene, 3 2h-thiophenone, dihydro, 3-tetrahydrothiophenone, tetrahydrothiophene-3-one, 3-oxo-2,3,4,5-tetrahydrothiophene
InChI Key DSXFPRKPFJRPIB-UHFFFAOYSA-N
SMILES O=C1CCSC1
IUPAC Name thiolan-3-one
Molecular Weight (g/mol) 102.15
PubChem CID 61252
Formula Weight 102.16
Percent Purity 98%
Chemical Name or Material Tetrahydrothiophen-3-one
Show More Show Less
compliance-icons
Product Identifier
  • Tetrahydrothiophen-3-one
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.