Learn More
Tetrahydrothiophene 1-oxide, 97%, Thermo Scientific Chemicals
Description
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
In a comparative study of aromatic fluorodenitration with Potassium fluoride, in several dipolar aprotic solvents, a much faster reaction rate was observed with DMSO or tetramethylene sulfoxide than with the more commonly used Sulfolane.
Solubility
Fully miscible in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.

Specifications
Specifications
Density | 1.173 |
Boiling Point | 235°C to 237°C |
Flash Point | 112°C (233°F) |
Refractive Index | 1.52 |
Quantity | 10 g |
Beilstein | 105274 |
Solubility Information | Fully miscible in water. |
Formula Weight | 104.17 |
Percent Purity | 97% |
Chemical Name or Material | Tetrahydrothiophene 1-oxide |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10240553 | Certificate of Analysis | 24/10/2023 | A17502.18,A17502.09 |
10240919 | Certificate of Analysis | 26/05/2022 | A17502.18,A17502.09 |
10195700 | Certificate of Analysis | 23/11/2021 | A17502.18,A17502.09 |
10179223 | Certificate of Analysis | 23/11/2021 | A17502.18,A17502.09 |
10153544 | Certificate of Analysis | 23/11/2021 | A17502.18,A17502.09 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.