missing translation for 'onlineSavingsMsg'
Learn More

Tetrakis(hydroxymethyl)phosphonium chloride, ca. 80% solution in water, Thermo Scientific Chemicals

42.91 € - 259.29 €

Specifications

Chemical Name or Material Tetrakis(hydroxymethyl)phosphonium chloride
CAS 124-64-1, 7732-18-5
Color Green-Yellow or Pink
Density 1.3400g/mL
Boiling Point 115.0°C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10350444
View Documents
Thermo Scientific Acros
389441000
100 mL Glass bottle
42.91 €
100mL
Please to purchase this item. Need a web account? Register with us today!
10045553
View Documents
Thermo Scientific Acros
389448000
800 mL Glass bottle
259.29 €
800mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for C-C Bond Formation (Olefination)
Specifications

Specifications

Tetrakis(hydroxymethyl)phosphonium chloride
Green-Yellow or Pink
115.0°C
Authentic
C4H12ClO4P
1.34
Solubility in water: soluble.
[Cl-].OC[P+](CO)(CO)CO
190.56
190.56
Solution
124-64-1, 7732-18-5
1.3400g/mL
96°C
Glass bottle
MFCD00031687
tetrakis hydroxymethyl phosphonium chloride, thpc, pyroset tkc, retardol c, tetramethylolphosphonium chloride, tetrakis hydroxymethyl phosphanium chloride, unii-58wb2xcf8i, proban cc, tetrakis hydroxymethyl phosphochloride, ccris 317
AKXUUJCMWZFYMV-UHFFFAOYSA-M
tetrakis(hydroxymethyl)phosphanium;chloride
31298
80.0 to 85.0%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May be corrosive to metals.
Toxic if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Suspected of damaging the unborn child.
Very toxic to aquatic life with lon

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 204-707-7

Product Identifier
  • Tetrakis(hydroxymethyl)phosphonium chloride

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Reproductive toxicity Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H361d-Suspected of damaging the unborn child.
  • H301-Toxic if swallowed.
  • H290-May be corrosive to metals.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only