Learn More
Tetrakis(trimethylsilyl)silane, 98%, Thermo Scientific Chemicals
37.89 € - 111.56 €
Chemical Identifiers
CAS | 4098-98-0 |
---|---|
Molecular Formula | C12H36Si5 |
Molecular Weight (g/mol) | 320.85 |
MDL Number | MFCD00054859 |
InChI Key | BOJSDHZZKKYWAS-UHFFFAOYSA-N |
Synonym | tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa |
PubChem CID | 138115 |
IUPAC Name | tetrakis(trimethylsilyl)silane |
SMILES | C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11490097
|
Thermo Scientific Alfa Aesar
B21020.03 |
1 g |
37.89 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11400107
|
Thermo Scientific Alfa Aesar
B21020.06 |
5 g |
111.56 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used as a starting material for the synthesis of tris(trimethylsilyl)silyl lithium with methyl lithium and in the preparation of lithium bis[(TMS)3silyl]cuprate.
Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Chemical Identifiers
4098-98-0 | |
320.85 | |
BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
138115 | |
C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C |
C12H36Si5 | |
MFCD00054859 | |
tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
tetrakis(trimethylsilyl)silane |
Specifications
266°C to 268°C (sublimation) | |
C12H36Si5 | |
tetrakis trimethylsilyl silane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl trisilane, tetrakil trimethylsilyl silane, si si ch3 3 4, trisilane, 1,1,1,3,3,3-hexamethyl-2,2-bis trimethylsilyl, 2,2-bis trimethylsilyl-1,1,1,3,3,3-hexamethyl-trisilane, acmc-1an6g, tetra trimethylsilyl-silane, bojsdhzzkkywas-uhfffaoysa | |
C[Si](C)(C)[Si]([Si](C)(C)C)([Si](C)(C)C)[Si](C)(C)C | |
320.85 | |
320.85 | |
Tetrakis(trimethylsilyl)silane |
4098-98-0 | |
MFCD00054859 | |
BOJSDHZZKKYWAS-UHFFFAOYSA-N | |
tetrakis(trimethylsilyl)silane | |
138115 | |
98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- Tetrakis(trimethylsilyl)silane
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Tetrakis(trimethylsilyl)silane
RUO – Research Use Only