missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetrakis(triphenylphosphine)palladium(0), 99.8% (metals basis), Pd 9% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
0.5 g
2 g
10 g
Description
Widely used as a catalyst for palladium-catalyzed coupling reactions including Suzuki coupling, Sonogashira coupling, Heck reaction, Negishi coupling and Stille coupling. Used in isomerization, carbonylation, oxidation, and C-C bond formation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 100°C to 105°C |
Assay Percent Range | 99.8% (metals basis), Pd 9% min |
Quantity | 10 g |
Beilstein | 6704828 |
Sensitivity | Air sensitive |
Synonym | tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4 |
Solubility Information | Insoluble in water. Soluble in benzene,ethanol and chloroform |
IUPAC Name | palladium;triphenylphosphane |
Formula Weight | 1155.57 |
Physical Form | Crystalline |
Show More |
Safety and Handling
Product Identifier
- Tetrakis(triphenylphosphine)palladium(0)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- MIXTURE LIST-Contains : Tetrakis(triphenylphosphine)palladium(0)
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'