missing translation for 'onlineSavingsMsg'
Learn More
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals Product Code.: 10492391

Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals

Product Code. 10492391
5 g, Glass bottle
Click to view available options
Quantity:
1 g
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10492391

Brand: Thermo Scientific Acros 202380050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14221-01-3
Molecular Formula C72H60P4Pd
Molecular Weight (g/mol) 1155.59
MDL Number MFCD00010012
InChI Key NFHFRUOZVGFOOS-UHFFFAOYSA-N
Synonym tetrakis triphenylphosphine palladium, tetrakis triphenylphosphine palladium 0, pd pph3 4, tetrakis triphenylphosphine palladium o, tetra triphenylphosphine palladium, palladium-tetrakis triphenylphosphine, palladium 0 tetrakis triphenylphosphine, palladium, tetrakis triphenylphosphine-, t-4
PubChem CID 11979704
IUPAC Name palladium;triphenylphosphane
SMILES [Pd].C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1.C1=CC=C(C=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 103.0°C to 107.0°C
Color Khaki to Yellow
Elemental Analysis 98.5% min.
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula [(C6H5)3P]4Pd
Quantity 5 g
Fieser 06,571; 07,357; 08,472; 09,451; 10,384; 11,503; 12,468; 13,34; 14,177; 16,98; 17,327
Merck Index 15,9367
Solubility Information Solubility in water: insoluble.
Formula Weight 1155.58
Percent Purity 99%
Physical Form Fine Powder or Platelets
Chemical Name or Material Tetrakis(triphenylphosphine)palladium(0)
Show More Show Less
compliance-icons
Product Identifier
  • Tetrakis(triphenylphosphine)palladium(0)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Tetrakis(triphenylphosphine)palladium(0), 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.