missing translation for 'onlineSavingsMsg'
Learn More

beta-Tetralone, 95%

Product Code. 10105700 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10105700

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 530-93-8 | C10H10O | 146.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 530-93-8
Molecular Formula C10H10O
Molecular Weight (g/mol) 146.19
MDL Number MFCD00001727
InChI Key KCKZIWSINLBROE-UHFFFAOYSA-N
Synonym 2-tetralone, beta-tetralone, 1,2,3,4-tetrahydro-2-naphthalenone, 3,4-dihydronaphthalen-2 1h-one, 1,2,3,4-tetrahydronaphthalen-2-one, 2 1h-naphthalenone, 3,4-dihydro, .beta.-tetralone, tetralin-2-one, 3,4-dihydro-2 1h-naphthalenone, 3,4-dihydro-2-oxonaphthalene
PubChem CID 68266
IUPAC Name 3,4-dihydro-1H-naphthalen-2-one
SMILES C1CC2=CC=CC=C2CC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 18.0°C
Color Yellow
Density 1.1060g/mL
Boiling Point 131.0°C to 133.0°C (11.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5590 to 1.5630
Quantity 5 g
Beilstein 07, 370
Specific Gravity 1.106
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether and benzene
Formula Weight 146.19
Percent Purity 95%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material β-Tetralone, 95%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.