missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals

29.75 € - 165.83 €

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 10% in Water
CAS 75-59-2, 7732-18-5
Color Colorless
Density 1.0000g/mL
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10167850
View Documents Promotion Details
Thermo Scientific Acros
420511000
100 g Plastic bottle
29.75 €
100g
Please to purchase this item. Need a web account? Register with us today!
10712032
View Documents Promotion Details
Thermo Scientific Acros
420515000
500 g Plastic bottle
106.53 €
500g
Please to purchase this item. Need a web account? Register with us today!
10742784
View Documents Promotion Details
Thermo Scientific Acros
420510010
1 kg Plastic bottle
165.83 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hydroxide
75-59-2, 7732-18-5
1.0000g/mL
Plastic bottle
(CH3)4NOH
04, 50
15, 9371
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
9.5 to 10.5%
10% in Water
Colorless
100.0°C
C4H13NO
MFCD00008280
11,514
1
Solubility in water: soluble.
C[N+](C)(C)C.[OH-]
91.15
91.15
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-882-9

missing translation for 'rtecsNumber' : PA0875000

missing translation for 'tsca' : TSCA

Product Identifier
  • Tetramethylammonium hydroxide, 10% in water

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 1
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
  • P331-Do NOT induce vomiting.
  • P363-Wash contaminated clothing before reuse.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Tetramethylammoniumhydroxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only