missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide, 25 wt% in methanol, Thermo Scientific Chemicals

127.64 € - 408.00 €

Specifications

Chemical Name or Material Tetramethylammonium hydroxide
Name Note 25 wt. % in Methanol
CAS 75-59-2, 67-56-1
Boiling Point 65.0°C
Melting Point -98.0°C
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10274850
View Documents
Thermo Scientific Acros
138351000
100 g Plastic bottle
176.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
10284850
View Documents
Thermo Scientific Acros
138355000
500 g Plastic bottle
127.64 €
500g
Please to purchase this item. Need a web account? Register with us today!
11975441
View Documents
Thermo Scientific Acros
138350025
2.5 kg Plastic Bottle
408.00 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Phase transfer catalyst
Specifications

Specifications

Tetramethylammonium hydroxide
75-59-2, 67-56-1
-98.0°C
(CH3)4NOH
0.9000g/mL
Solubility in water: soluble.
C[N+](C)(C)C.[OH-]
91.154
91.15
6°C
25 wt. % in Methanol
65.0°C
C4H13NO
MFCD00008280
tetramethylammonium hydroxide, tmah, hydroxyde de tetramethylammonium, nmw-w, nmd 3, tetramethyl ammonium hydroxide, unii-5gkp7317q2, ammonium, tetramethyl-, hydroxide, methanaminium, n,n,n-trimethyl-, hydroxide, tetramethylammoniumhydroxide
WGTYBPLFGIVFAS-UHFFFAOYSA-M
tetramethylazanium;hydroxide
60966
0.9
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Toxic in contact with skin.
Toxic if swallowed.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

missing translation for 'einecsNumber' : 200-882-9

missing translation for 'rtecsNumber' : PA0875000

missing translation for 'tsca' : TSCA

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 1
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H331-Toxic if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H370-Causes damage to organs.
  • H310-Fatal in contact with skin.
  • H372-Causes damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P350-IF ON SKIN: Gently wash with plenty of soap and water.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

Supplemental information
  • MIXTURE LIST-Contains : Methyl alcohol, Tetramethylammonium hydroxide

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.