missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium hydroxide pentahydrate, 98%, Thermo Scientific Chemicals

Product Code. 11332187
Click to view available options
Quantity:
25 g
100 g

Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Tetramethylammonium hydroxide pentahydrate is used in analytical chemistry as an ion pairing agent and a polarographic reagent. It finds application in gas chromatography/mass spectrometry (GC/MS), capillary isotachophoresis and capillary zone electrophoresis and liquid chromatography. Furthermore, it is utilized in the preparation of iron oxide nanoparticles which are useful as magnetic resonance imaging contrast agents and tetramethylammonium dithiocarboxylates. It acts as a developer or an etchant in semiconductor and photoelectric industries.

Solubility
Soluble in water and methanol.

Notes
Air sensitive. Hygroscopic. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10424-65-4
Molecular Formula C4H23NO6
Molecular Weight (g/mol) 181.23
MDL Number MFCD00149566
InChI Key MYXKPFMQWULLOH-UHFFFAOYSA-M
Synonym tetramethylammonium hydroxide pentahydrate, b2hkt2lckq, unii-b2hkt2lckq, methanaminium, n,n,n-trimethyl-, hydroxide, pentahydrate, tmaoh, tetramethylammoniumhydroxid, tetrametanoamonio hydroxido, tetrametiloamonio hydroxido, methanaminium, n,n,n-trimethyl-, hydroxide, hydrate 1:1:5, acmc-2098bm
PubChem CID 82620
IUPAC Name tetramethylazanium;hydroxide;pentahydrate
SMILES O.O.O.O.O.[OH-].C[N+](C)(C)C

Specifications

Melting Point 66°C to 67°C
Linear Formula (CH3)4NOH·5H2O
Quantity 25 g
UN Number UN3423
Beilstein 3714235
Sensitivity Air sensitive; Hygroscopic
Merck Index 14,9224
Solubility Information Soluble in water and methanol.
Formula Weight 181.23 (91.15 Anhydrous)
Percent Purity 98%
Chemical Name or Material Tetramethylammonium hydroxide pentahydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'