missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylammonium perchlorate, 98%, Thermo Scientific Chemicals

Product Code. 11955451
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.
This item is not returnable. View return policy
Sea freight only: For EU stock allow up to 15 days delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent, Phase transfer catalyst
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2537-36-2
Molecular Formula C4H12ClNO4
Molecular Weight (g/mol) 173.59
MDL Number MFCD00043207
InChI Key ZCWKIFAQRXNZCH-UHFFFAOYSA-M
Synonym tetramethylammonium perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate, n,n,n-trimethylmethanaminium perchlorate, ammonium, tetramethyl-, perchlorate, methanaminium, n,n,n-trimethyl-, perchlorate 1:1, c4h12n.clo4, acmc-20ajj6, tetramethylazanium perchlorate, tetramethylammonium perchlorate 25g, wln: 2k2&2&2 &wgw
PubChem CID 17337
IUPAC Name tetramethylazanium;perchlorate
SMILES C[N+](C)(C)C.[O-][Cl](=O)(=O)=O

Specifications

Melting Point 300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (on dry base)
Packaging Plastic Bottle
Linear Formula (CH3)4NClO4
Quantity 100 g
Formula Weight 173.6
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetramethylammonium perchlorate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Tetramethylammonium perchlorate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Oxidising solid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H272-May intensify fire; oxidiser.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'