missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tetramethylene sulfoxide, 97%, Thermo Scientific Chemicals
27.00 € - 210.00 €
Chemical Identifiers
CAS | 1600-44-8 |
---|---|
Molecular Formula | C4H8OS |
Molecular Weight (g/mol) | 104.17 |
MDL Number | MFCD00005477 |
InChI Key | ISXOBTBCNRIIQO-UHFFFAOYSA-N |
Synonym | tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide |
PubChem CID | 1128 |
IUPAC Name | thiolane 1-oxide |
SMILES | C1CCS(=O)C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10260972
|
Thermo Scientific Acros
138440050 |
5 g | Glass bottle |
27.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10306500
|
Thermo Scientific Acros
138440250 |
25 g | Glass bottle |
73.80 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10285040
|
Thermo Scientific Acros
138441000 |
100 g | Glass bottle |
210.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1600-44-8 | |
104.17 | |
ISXOBTBCNRIIQO-UHFFFAOYSA-N | |
1128 | |
C1CCS(=O)C1 |
C4H8OS | |
MFCD00005477 | |
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide | |
thiolane 1-oxide |
Specifications
1600-44-8 | |
1.1500g/mL | |
>112°C | |
96% min. (GC) | |
1.5180 to 1.5220 | |
01,1145 | |
tetramethylene sulfoxide, tetrahydrothiophene 1-oxide, thiophane oxide, tetrahydrothiophene oxide, thiophane monoxide, thiophane 1-oxide, thiophene, tetrahydro-, 1-oxide, tetramethylene sulphoxide, thiophene, tetrahydro, 1-oxide, tetrametylene sulfoxide | |
C1CCS(=O)C1 | |
104.17 | |
104.17 | |
Liquid |
Colorless to Yellow | |
235.0°C to 237.0°C | |
Authentic | |
C4H8OS | |
MFCD00005477 | |
1.15 | |
ISXOBTBCNRIIQO-UHFFFAOYSA-N | |
thiolane 1-oxide | |
1128 | |
97% | |
Tetramethylene sulfoxide, 98+% |
Safety and Handling
missing translation for 'einecsNumber' : 216-493-2
missing translation for 'rtecsNumber' : XN0830000
missing translation for 'tsca' : TSCA
RUO – Research Use Only