missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Tetramethylrhodamine-5-Iodoacetamide Dihydroiodide (5-TMRIA), single isomer

Tetramethylrhodamine-5-Iodoacetamide Dihydroiodide (5-TMRIA), single isomer

Brand:  Molecular Probes™ T6006

568.48 EUR valid until 2025-03-28
Use promo code "24107" to get your promotional price.


Product Code. 11550786

  • 768.00 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The thiol-reactive tetramethylrhodamine-5-iodoacetamide dihydroiodide (5-TMRIA) can be used to can be used to create bright orange-red-fluorescent bioconjugates with excitation/emission maxima ∼555/580.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Rhodamine Isomers, TMR (Tetramethylrhodamine)
580
Tetramethylrhodamine-5-Iodoacetamide Dihydroiodide
Thiol
Alkyl Halide, Iodoacetamide
Classic Dyes
555
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2903460 Certificate TypeCertificate of Analysis Date26/05/2024 Catalog NumberT6006
Lot Number2816165 Certificate TypeCertificate of Analysis Date02/12/2023 Catalog NumberT6006
Lot Number2425167 Certificate TypeCertificate of Analysis Date26/01/2023 Catalog NumberT6006
Lot Number2417137 Certificate TypeCertificate of Analysis Date14/10/2021 Catalog NumberT6006
Lot Number2413131 Certificate TypeCertificate of Analysis Date13/09/2021 Catalog NumberT6006
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molecular Probes™ Tetramethylrhodamine-5-Iodoacetamide Dihydroiodide (5-TMRIA), single isomer > 5mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.