Learn More
Tetramethylsilane, 99.9%, Thermo Scientific Chemicals
39.00 € - 99.80 €
Chemical Identifiers
CAS | 75-76-3 |
---|---|
Molecular Formula | C4H12Si |
Molecular Weight (g/mol) | 88.23 |
MDL Number | MFCD00008274 |
InChI Key | CZDYPVPMEAXLPK-UHFFFAOYSA-N |
Synonym | silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 |
PubChem CID | 6396 |
ChEBI | CHEBI:85361 |
IUPAC Name | tetramethylsilane |
SMILES | C[Si](C)(C)C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11484524
|
Thermo Scientific Alfa Aesar
A13148.14 |
25 g |
39.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494524
|
Thermo Scientific Alfa Aesar
A13148.22 |
100 g |
99.80 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsTetramethylsilane is used as a building block in organometallic chemistry. It acts as a by-product in the production of methyl chlorosilanes. Also, it serves as a precursor to silicon dioxide or silicon carbide. It is used as internal reference standard for the calibration of chemical sift for 1, 13 and 29 NMR spectroscopy. In addition, it is used as an aviation fuel.
Solubility
Miscible with water, ethanol, ether and most organic solvents. Immiscible with cold sulfuric acid.
Notes
Store in a cool place. Incompatible with strong acids, strong bases and strong oxidizing agents.
Chemical Identifiers
75-76-3 | |
88.23 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
6396 | |
tetramethylsilane |
C4H12Si | |
MFCD00008274 | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CHEBI:85361 | |
C[Si](C)(C)C |
Specifications
26°C to 27°C | |
75-76-3 | |
MFCD00008274 | |
1696908 | |
14,9229 | |
Miscible with water,ethanol,ether and most organic solvents. Immiscible with cold sulfuric acid. | |
C[Si](C)(C)C | |
88.23 | |
CHEBI:85361 | |
99.9% | |
1.359 | |
Tetramethylsilane |
-100°C | |
C4H12Si | |
UN2749 | |
−27°C (−16°F) | |
silane, tetramethyl, tetramethylsilicane, tetramethyl silane, silicon, tetramethyl, tetramethyl-silane, unii-41y0rbg14q, me4si, ch3 4si, si ch3 4, chembl68073 | |
CZDYPVPMEAXLPK-UHFFFAOYSA-N | |
tetramethylsilane | |
6396 | |
88.23 | |
Mild | |
0.646 |
Safety and Handling
GHS H Statement
H224
Extremely flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H224
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: I; Proper Shipping Name: TETRAMETHYLSILANE
missing translation for 'einecsNumber' : 200-899-1
missing translation for 'rtecsNumber' : VV5706000
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold
- Danger
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Flammable liquid Category 1
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
RUO – Research Use Only