missing translation for 'onlineSavingsMsg'
Learn More

Tetramethylthiuram monosulfide, 97%, Thermo Scientific Chemicals

Product Code. 11443487
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-74-5
Molecular Formula C6H12N2S3
Molecular Weight (g/mol) 208.356
MDL Number MFCD00014870
InChI Key REQPQFUJGGOFQL-UHFFFAOYSA-N
Synonym tetramethylthiuram monosulfide, bis dimethylthiocarbamoyl sulfide, monothiuram, unads, mono-thiurad, tmtm, thiuram mm, aceto tmtm, cyuram ms, ekagom tm
PubChem CID 7347
IUPAC Name dimethylcarbamothioyl N,N-dimethylcarbamodithioate
SMILES CN(C)C(=S)SC(=S)N(C)C

Specifications

Melting Point 106°C to 110°C
Density 1.37
Quantity 500 g
Beilstein 1775650
Formula Weight 208.37
Percent Purity 97%
Chemical Name or Material Tetramethylthiuram monosulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'