Learn More
Thermo Scientific Chemicals Tetranitro blue tetrazolium chloride, 97%
Redox indicator for enzymes
82.51 € - 519.59 €
Chemical Identifiers
CAS | 1184-43-6 |
---|---|
Molecular Formula | C40H28Cl2N12O10 |
Molecular Weight (g/mol) | 907.64 |
MDL Number | MFCD00036338 |
InChI Key | VCESGVLABVSDRO-UHFFFAOYSA-L |
Synonym | TNBT |
PubChem CID | 129893520 |
SMILES | [Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15453739
|
Thermo Scientific Chemicals
J61080.MC |
100 mg |
82.51 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15443739
|
Thermo Scientific Chemicals
J61080.03 |
1 g |
519.59 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Redox indicator for enzymes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1184-43-6 | |
907.64 | |
VCESGVLABVSDRO-UHFFFAOYSA-L | |
129893520 |
C40H28Cl2N12O10 | |
MFCD00036338 | |
TNBT | |
[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O |
Specifications
1184-43-6 | |
MFCD00036338 | |
TNBT | |
[Cl-].[Cl-].COC1=CC(=CC=C1[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C(OC)=C1)[N+]1=NC(=NN1C1=CC=C(C=C1)[N+]([O-])=O)C1=CC=C(C=C1)[N+]([O-])=O | |
907.64 | |
999.78 | |
Yellow | |
Tetranitro blue tetrazolium chloride |
C40H28Cl2N12O10 | |
3897475 | |
VCESGVLABVSDRO-UHFFFAOYSA-L | |
2-{4'-[3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium-2-yl]-3,3'-dimethoxy-[1,1'-biphenyl]-4-yl}-3,5-bis(4-nitrophenyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium dichloride | |
129893520 | |
97% | |
Powder |
Safety and Handling
GHS H Statement
H350
May cause cancer.
P201-P202-P281-P308+P313-P501c
H350
missing translation for 'einecsNumber' : 214-665-1
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.