missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenylcyclopentadienone, 99%, Thermo Scientific Chemicals

Product Code. 10234710
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 479-33-4
Molecular Formula C29H20O
Molecular Weight (g/mol) 384.47
MDL Number MFCD00001407
InChI Key PLGPSDNOLCVGSS-UHFFFAOYSA-N
Synonym tetraphenylcyclopentadienone, tetracyclone, cyclone, tetracyclon, 2,3,4,5-tetraphenylcyclopenta-2,4-dienone, 2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenyl, tetraphenyl-2,4-cyclopentadien-1-one, 2,3,4,5-tetraphenylcyclopentadienone, cyclone compound, 2,3,4,5-tetraphenyl-2,4-cyclopentadienone
PubChem CID 68068
IUPAC Name 2,3,4,5-tetraphenylcyclopenta-2,4-dien-1-one
SMILES C1=CC=C(C=C1)C2=C(C(=O)C(=C2C3=CC=CC=C3)C4=CC=CC=C4)C5=CC=CC=C5

Specifications

Melting Point 217.0°C to 222.0°C
Color Black
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 100 g
Fieser 01,1149; 04,490; 06,577
Solubility Information Solubility in water: insoluble.
Formula Weight 384.47
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material Tetraphenylcyclopentadienone, 99%
Show More Show Less
Product Identifier
  • Tetraphenylcyclopentadienone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'