missing translation for 'onlineSavingsMsg'
Learn More

Tetraphenyllead, 97%, Thermo Scientific™

Product Code. 11414883
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 595-89-1
Molecular Formula C24H20Pb
Molecular Weight (g/mol) 515.60
MDL Number MFCD00002999
InChI Key WBJSMHDYLOJVKC-UHFFFAOYSA-N
Synonym tetraphenyllead, tetraphenyl lead, plumbane, tetraphenyl, pbph4, lead, tetraphenyl-,, lead tetraphenyl, acmc-209mel, wbjsmhdylojvkc-uhfffaoysa
PubChem CID 72906
ChEBI CHEBI:30184
IUPAC Name tetraphenylplumbane
SMILES C1=CC=C(C=C1)[Pb](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 227°C to 228°C
Boiling Point 126°C (13 mmHg)
Quantity 5 g
UN Number UN3467
Formula Weight 515.61
Percent Purity 97%
Chemical Name or Material Tetraphenyllead
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Tetraphenyllead
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.