Learn More
Tetrapropylammonium Hydroxide, 10% in Water, Thermo Scientific Chemicals
56.40 € - 194.00 €
Chemical Identifiers
CAS | 4499-86-9, 7732-18-5 |
---|---|
Molecular Formula | C12H29NO |
Molecular Weight (g/mol) | 203.37 |
MDL Number | MFCD00009360 |
InChI Key | LPSKDVINWQNWFE-UHFFFAOYSA-M |
Synonym | tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide |
PubChem CID | 20586 |
IUPAC Name | tetrapropylazanium hydroxide |
SMILES | [OH-].CCC[N+](CCC)(CCC)CCC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10731642
|
Thermo Scientific Acros
420721000 |
100 g | Plastic bottle |
56.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10065960
|
Thermo Scientific Acros
420725000 |
500 g | Plastic bottle |
194.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalystChemical Identifiers
4499-86-9, 7732-18-5 | |
203.37 | |
LPSKDVINWQNWFE-UHFFFAOYSA-M | |
20586 | |
[OH-].CCC[N+](CCC)(CCC)CCC |
C12H29NO | |
MFCD00009360 | |
tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide | |
tetrapropylazanium hydroxide |
Specifications
Colorless | |
100.0°C to 102.0°C | |
(CH3CH2CH2)4NOH | |
1.0000g/mL | |
Solubility in water: soluble in water. | |
[OH-].CCC[N+](CCC)(CCC)CCC | |
203.37 | |
203.37 | |
1 | |
Tetrapropylammonium hydroxide, 10% in Water |
4499-86-9, 7732-18-5 | |
C12H29NO | |
MFCD00009360 | |
tetrapropylammonium hydroxide, tetra-n-propylammonium hydroxide, 1-propanaminium, n,n,n-tripropyl-, hydroxide, tpaoh, tetrapropylammonium oxide, unii-t686luy7nk, tetrapropylazanium hydroxide, t686luy7nk, ammonium, tetrapropyl-, hydroxide, n,n,n-tripropyl-1-propanaminium hydroxide | |
LPSKDVINWQNWFE-UHFFFAOYSA-M | |
tetrapropylazanium hydroxide | |
20586 | |
9 to 11% | |
Liquid |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 224-800-6
missing translation for 'rtecsNumber' : BS8400000
missing translation for 'tsca' : TSCA
RUO – Research Use Only