Learn More
Tetrathiafulvalene, 97%, Thermo Scientific Chemicals
Description
Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Tetrathiafulvalene, holds wide application in HPLC, NMR. This heterocyclic compound contributed to the development of molecular electronics. They are used as a organic super conductors.
Solubility
It is insoluble in water. Soluble in organic solvents.
Notes
Air & Light Sensitive. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.

Specifications
Specifications
Melting Point | 116°C to 120°C |
Quantity | 250 mg |
Beilstein | 1617956 |
Sensitivity | Air and light sensitive |
Merck Index | 14,9242 |
Solubility Information | It is insoluble in water. Soluble in organic solvents. |
Formula Weight | 204.35 |
Percent Purity | 97% |
Chemical Name or Material | Tetrathiafulvalene |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10251764 | Certificate of Analysis | 30/08/2024 | L19229.03,L19229.MD |
10241640 | Certificate of Analysis | 29/06/2022 | L19229.03,L19229.MD |
F13X073 | Certificate of Analysis | 08/12/2021 | L19229.03,L19229.MD |
F24Z55 | Certificate of Analysis | 08/12/2021 | L19229.03,L19229.MD |
10153738 | Certificate of Analysis | 08/12/2021 | L19229.03,L19229.MD |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.