missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Tetrazolium Violet, 98+%

Product Code. 11498616
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11498616

Brand: Thermo Scientific Chemicals A16008.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1719-71-7 | C23H17ClN4 | 384.867 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1719-71-7
Molecular Formula C23H17ClN4
Molecular Weight (g/mol) 384.867
MDL Number MFCD00011875
InChI Key RONADMZTCCPLEF-UHFFFAOYSA-M
Synonym tetrazolium violet, violet tetrazolium, tetrazolium purple, tv, 2,5-diphenyl-3 1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-1-naphthyl-2h-tetrazolium chloride, 2,5-diphenyl-3-alpha-naphthyl-2h-tetrazolium chloride, 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium chloride, 2h-tetrazolium, 3-1-naphthyl-2,5-diphenyl-, chloride, tetrazolium violet at
PubChem CID 74395
ChEBI CHEBI:75193
IUPAC Name 2-naphthalen-1-yl-3,5-diphenyltetrazol-2-ium;chloride
SMILES C1=CC=C(C=C1)C2=NN([N+](=N2)C3=CC=CC4=CC=CC=C43)C5=CC=CC=C5.[Cl-]

Specifications

Melting Point 247°C to 249°C
Quantity 25 g
Formula Weight 384.87
Percent Purity ≥98%
Chemical Name or Material Tetrazolium Violet

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt