missing translation for 'onlineSavingsMsg'
Learn More

Texas Instruments Graphing Calculator with Advanced Statistics

The Graphing Calculator with Advanced Statistics features seven graphing styles to differentiate graphs, an equation solver editor, and flash technology that enables you to customize your calculator.

Brand:  Texas Instruments 17311-12

Product Code. 11720209

  • 255.00 € / Each
Estimated Shipment: 30-06-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Seven graphing styles differentiate graphs: line, thick-line, dotted-line, shading, and more.
  • Equation solver editor solves for different variables interactively.
  • Advanced statistics: hypothesis testing, confidence intervals, and distribution.
  • Store groups of related data in the archive for later use.
  • Regression analysis with sine and logistic regressions.
  • Flash technology enables you to customize your calculator by downloading software (requires TI-GRAPH LINK™).
  • What's included: link cable (to connect to other calculators), four AAA batteries, slide case, and instructions.
Specifications

Specifications

Graphing Calculator
63.5 W x 38.1 H mm
24 KB RAM, 160 KB Data Storage
1 Ea.
182.5625 mm
88.9 mm
Four AAA batteries
LCD
Advanced Statistics: Hypothesis Testing, Confidence Intervals, and Distribution
TI-83+
7.2 in.
3.5 in.
AAA
Link Cable, Four AAA Batteries, Slide Case, and Instructions
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Texas Instruments Graphing Calculator with Advanced Statistics > Graphing Calculator with Advanced Statistics; Display: LCD

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.