missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ Texas Red™ Sulfonyl Chloride, mixed isomers
Texas Red™ Sulfonyl Chloride, mixed isomers
Brand: Molecular Probes™ T1905
410.40 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The amine-reactive Texas Red™ sulfonyl chloride can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.Specifications
10 x 1 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
Texas Red™ | |
Texas Red™ | |
595 |
Sulfonyl Chloride | |
Amine | |
Sulfonyl Chloride | |
Classic Dyes | |
615 |
For Research Use Only. Not for use in diagnostic procedures.