missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Texas Red™ Sulfonyl Chloride, mixed isomers

Texas Red™ Sulfonyl Chloride, mixed isomers

Brand:  Molecular Probes™ T353

335.92 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11580776

  • 442.00 € / 10mg
Estimated Shipment: 06-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

The amine-reactive Texas Red™ sulfonyl chloride can be used to can be used to create bright red-fluorescent bioconjugates with excitation/emission maxima ∼595/615 nm.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

10 mg
Store in freezer (-5 to -30°C) and protect from light.
Room Temperature
Texas Red™
Texas Red™
595
Sulfonyl Chloride
Amine
Sulfonyl Chloride
Classic Dyes
615
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.