Learn More
Texwipe™ Alpha™ 10 Sealed-Border Wipers
For ISO Class 3 to 4 applications
Brand:  Texwipe™ TX3280
 View more versions of this product
Product Code. 15604987
  • 955.00 € / Pack of 250
Explore more special offers
Description
Description
  • Vectra™ processed—hands-free, continuous manufacturing purification and inspection
  • Double-knit, no-run interlock construction of 100% continuous-filament polyester
  • Patented sealed-border wiper for lowest releasable fibers and particles
  • Optimum balance of wiping efficiency, abrasion resistance, and sorbency
  • Ideal wiper for use in multiple environments
  • Lot-to-lot traceability
Order Info
Available dry or sterile and presaturated with isopropanol. Basis weight: 130g/m2. Sterile wipers 19-130-2061 packed 50 wipers/ flatpack, 5 flatpacks /case; 19-165-653 packed 20 wipers per flexpack, 20 flexpacks/case.
Specifications
Specifications
Pre-wetted Wipe
30.5 cm
30.5 cm
ISO 9001:2000, 13485:2003, 14001:2004 and OHSAS 18001:1999
12 x 12 in. (30.48 x 30.48 cm)
70% Isopropyl Alcohol, 30% WFI
12 in.
12 in.
Polyester
Sealed
Ideal wipes for use in multiple environments
Sterile