Learn More
Texwipe™ AlphaMop™ Isolator Cleaning Tool
Excellent for cleaning isolators, laminar flow hoods, biosafety cabinets, and glove boxes
230.00 €
Specifications
Autoclavable Autoclavable
View More Specs
Products 1
Product Code Brand Material Product Type Includes Price Quantity & Availability  
Product Code Brand Material Product Type Includes Price Quantity & Availability  
11551214
View Documents
Texwipe™
TX7101
Plastic Mop
230.00 €
Each
Estimated Shipment: 31-03-2025
to see stock.
Add to basket
Description
Description
  • Low-profile mop with rounded corners and swivel head to reach difficult-to-access areas
  • Made from autoclavable Celcon™ plastic
  • Replaceable foam pad ensures that mop cover conforms to surfaces being cleaned
Specifications
Specifications
Autoclavable