Learn More
Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer
Apply directly to floor
Brand:  Texwipe™ AMA364681B
 View more versions of this product
Product Code. 11525325
  • 518.00 € / Pack of 8
Estimated Shipment: 05-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Apply directly to floor
  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area
Specifications
Specifications
46 x 36 in.
30 Sheets/Mat
Blue