missing translation for 'onlineSavingsMsg'
Learn More

Texwipe™ IdealMat™ Contamination Control Mats - 30 Layer

Apply directly to floor

257.00 € - 518.00 €

Specifications

Quantity 30 Sheets/Mat
View More Specs

Products 5
Product Code Brand Color Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Color Dimensions (L x W) Price Quantity & Availability  
11565315
View Documents
Texwipe™
AMA254581W
White 45 x 25 in.
337.00 €
Pack of 8
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
 
11525325
View Documents
Texwipe™
AMA364681B
Blue 46 x 36 in.
518.00 €
Pack of 8
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
 
11505325
View Documents
Texwipe™
AMA254581B
Blue 45 x 25 in.
337.00 €
Pack of 8
Estimated Shipment: 06-03-2025
to see stock.
Add to basket
 
12410070
View Documents
Texwipe™
AMA364681W
White 46 x 36 in.
518.00 €
Pack of 8
Estimated Shipment: 27-02-2025
to see stock.
Add to basket
 
15762386
View Documents
Texwipe™
AMA254581G
Gray 25 x 45 in. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

Apply directly to floor

  • Consistent adhesive coating on each layer captures particulates from foot traffic and equipment wheels
  • Antimicrobial agent in coating reduces potential for spreading germs and bacteria into controlled area

Specifications

Specifications

30 Sheets/Mat
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.