missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texwipe™ TechniSat™ Gowning Room Wipers
For ISO Class 6 applications
Brand: Texwipe™ TX1048
This item is not returnable.
View return policy
Description
- Nonwoven cellulose/polyester wipers presaturated with 6% semiconductor-grade isopropyl alcohol/94% deionized water solution
- Ideal for wiping items such as badges, pagers, cell phones, palmtop computers, notebooks, goggles and gloves in the gowning room area
- Help to reinforce contamination control protocols while reducing costs
- Designed for easy dispensing on demand to minimize contamination and eliminate the need to maintain bulk solvents and/or spray bottles
- Reusable flip-top tub is available for wiper storage and dispensing
Order Info
Packs are stacked and folded. Each case includes 12 packs of wipers and one flip-top tub. Additional flip-top tubs available separately.
Specifications
Pre-wetted Wipe | |
20.32 cm | |
12.7 cm | |
8 x 5 in. (20.32 x 12.7 cm) | |
6% Semiconductor grade Isopropyl Alcohol or 94% Deionized Water |
8 in. | |
5 in. | |
Polyester Cellulose | |
Designed for easy dispensing on demand to minimize contamination and eliminate the need to maintain bulk solvents and/or spray bottles |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- texwipes pre-wetted with 6% IPA
Signal Word
- Warning
Hazard Category
- Flammable solid Category 1
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H228-Flammable solid.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : isopropyl alcohol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction