Learn More
Texwipe™ TOC Cleaning Validation Kits
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Brand:  Texwipe™ TX3342
 View more versions of this product
Product Code. 10514495
  • 555.00 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Includes: Vials, swabs, and labels for sampling 12 or 72 production areas, and container for sample transport.
Description
Description
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
  • Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
  • Prewashed vials have one-piece cap and septa design for easy sampling and analysis
  • Polyethylene overcap protects septum surface from contamination until time of analysis
  • Sloped shoulder design minimizes headspace for reduced background TOC level
  • Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
Sampling Swab
  • Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
  • Double layered, double knit polyester head entraps contaminants in knit structure during sampling
  • Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
  • Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
  • 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Certifications
Certificate of Analysis included on each box.
Specifications
Specifications
Validation Kit
Sampling 72 Production Areas
72 40 mL clear vials bonded septa cap, 144 Low TOC Alpha™ Sampling Swab, 72 blank vial labels
1/Cs.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Texwipe™ TOC Cleaning Validation Kits > 72 production areas; 144 Low TOC Alpha Sampling Swab