missing translation for 'onlineSavingsMsg'
Learn More

Ebro Electronic™ TFI 550 Infrared Thermometer

Product Code. 10195614
Click to view available options
Temperature Range (Metric):
-60°C to +550°C
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10195614

Brand: Ebro Electronic™ 13401786A

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Infrared thermometer with thermocouple

Sinter filter/stainless steel for hygrometer
TRUSTED_SUSTAINABILITY

Specifications

Calibration Points 3 Points
Certifications/Compliance EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012
Dimensions (L x W x H) 46 x 121 x 195 mm
Emissivity 0.1 to 1.0
For Use With (Application) Infrared Dual Thermometer with connection for thermo elements type K for quick non-contact surface temperature measurement to use in incomming goods for instance
Height (Metric) 50 mm
Includes Thermometer, calibration certificate, instruction manual
Length (Metric) 190 mm
Material Plastic Housing
Model TFI 550
Resolution 0.1°C at &minus9.9°C to 199.9°C otherwise 1°C
Weight (Metric) 180 g
Product Type Infrared-Thermometer
Max. Temperature (English) 1022°F
Max. Temperature (Metric) 550°C
Min. Temperature (English) -76°F
Min. Temperature (Metric) -60°C
Temperature Range (Metric) -60°C to +550°C
Calibrated Yes
For Use With (Equipment) AN 141, AN 144, TPN probe series with SMP connector
Battery Life Approx. 140 hr.
Battery Type 2x AAA, 1.5 V
Accuracy ±2°C, -18°C to +23°C ±1%
Display LCD
Width (Metric) 153 mm
IP Rating IP20
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.