missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TGFBR3 Split FISH Probe
Labeled FISH probes for identification of gene split using Fluoresecent In Situ Hybridization Technique.
1515.00 € - 2495.00 €
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | transforming growth factor, beta receptor III |
Gene Alias | BGCAN,betaglycan |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16083485
|
Abnova™
FS0051.100uL |
100 μL |
1515.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16093485
|
Abnova™
FS0051.200uL |
200 μL |
2495.00 €
200µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
BGCAN,betaglycan | |
7049 | |
Liquid |
Tube | |
transforming growth factor, beta receptor III | |
TGFBR3 | |
DNA/RNA |