missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TH 10785

OGG1 activator

270.00 € - 1070.00 €

Specifications

Chemical Name or Material N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine
CAS 1002801-51-5
Target Glycosylase Activators
Molecular Formula C17H21N3
Purity 0.98
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18746956
View Documents
Tocris Bioscience™
7778/10
10 mg
270.00 €
10mg
Please to purchase this item. Need a web account? Register with us today!
18797406
View Documents
Tocris Bioscience™
7778/50
50 mg
1070.00 €
50mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TH 10785 is an 8-oxo guanine DNA glycosylase 1 (OGG1) activator. It binds in the active site of OGG1 and increases its recruitment to oxidative DNA damage and its repair.
Specifications

Specifications

N-Cyclohexyl-2-cyclopropyl-4-quinazolinamine
Glycosylase Activators
0.98
1002801-51-5
C17H21N3
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.