missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ TH-660 Swinging Bucket Rotor Package

Process samples effectively with the TH-660 Swinging Bucket Rotor.

Brand:  Thermo Scientific™ 10502

22084.00 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 12181630

  • 27605.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Process samples effectively with the Thermo Scientific™ TH-660 Swinging Bucket Rotor Package.

Ordering Information:

Order Re-Seal 11 × 32mm tube vise (45537) separately. Contact your Sales Representative for information.

Compatible with:

Thermo Scientific Sorvall WX80/90/100 Ultra Centrifuge Series

Recommended for:

Rate zonal isolation of proteins and RNA.

Note:

Conical Adapter Removal Tool (Parts only available directly from Seton Scientific) can be ordered as 4209

TRUSTED_SUSTAINABILITY
Specifications

Specifications

TH-660 Swinging Bucket Rotor Package
Swinging Bucket Rotor
488,576 x g
121.5 mm
44.4
11 mm
Thermo Scientific™ Sorvall™ WX ultraCentrifuges
374,173 x g
Titanium
6 x 4.4 mL
60,000 rpm
64.6 mm
60 mm
12.8 lb. (5.8 kg)
90°
259,770 x g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.