missing translation for 'onlineSavingsMsg'
Learn More

Thallic acetate, 98%, Thermo Scientific™

Product Code. 10381781
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 2570-63-0
Molecular Formula C6H9O6Tl
Molecular Weight (g/mol) 381.51
MDL Number MFCD00008698
Synonym Acetic acid, thallium(III) salt

Specifications

Melting Point 182°C
Color White
Physical Form Crystalline Powder
Quantity 25 g
Assay Percent Range 98%
Linear Formula (CH3CO2)3Tl
Beilstein 02, 96
Fieser 01,1150; 02,406; 03,286; 04,492; 05,655; 07,360
Merck Index 15, 9406
Solubility Information Solubility in water: decomposes
Formula Weight 381.51
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Thallic acetate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'