missing translation for 'onlineSavingsMsg'
Learn More

Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water, Thermo Scientific Chemicals

275.37 € - 960.78 €

Chemical Identifiers

CAS 7440-28-0
Molecular Formula Tl
Molecular Weight (g/mol) 204.38
MDL Number MFCD00134063
InChI Key BKVIYDNLLOSFOA-UHFFFAOYSA-N
Synonym ramor, thallium, metallic, thallium, elemental, unii-ad84r52xlf, thallium, soluble compounds, ad84r52xlf, thallium, elemental and soluble compounds, rod, 12.7mm 0.5in dia, granules, thallium
PubChem CID 5359464
ChEBI CHEBI:30440
IUPAC Name thallium
SMILES [Tl]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11326498
SDS View Documents
Thermo Scientific Alfa Aesar
012131.14
25 g
275.37 €
25g
Please to purchase this item. Need a web account? Register with us today!
11336498
SDS View Documents
Thermo Scientific Alfa Aesar
012131.22
100 g
960.78 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in water
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 231-138-1

missing translation for 'rtecsNumber' : XG3425000

missing translation for 'tsca' : Yes

missing translation for 'storageNote' :

  • Packaged in water

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium granules, 6mm (0.2in) & down, 99.99% (metals basis), packaged in water, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.