missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) bromide, 99.999% (metals basis)

Product Code. 11321908
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11321908

Brand: Thermo Scientific Alfa Aesar 011841.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7789-40-4

Thallium(I) bromide is used as Chemical reagents and pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7789-40-4
MDL Number MFCD00011273
Synonym Thallous bromide

Specifications

Melting Point ∼460°C
Physical Form Powder
Quantity 100 g
UN Number UN1707
Merck Index 14,9258
Solubility Information Practically insoluble in water
Formula Weight 284.28
Percent Purity 99.999%
Sensitivity Hygroscopic
Density 7.5 g/mL
Assay (metals basis)
Chemical Name or Material Thallium(I) bromide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.