Learn More
Thallium(I) chloride, ultra dry, 99.999% (metals basis), Thermo Scientific Chemicals
184.92 € - 622.10 €
Chemical Identifiers
CAS | 7791-12-0 |
---|---|
Molecular Formula | ClTl |
Molecular Weight (g/mol) | 239.83 |
MDL Number | MFCD00011274 |
InChI Key | GBECUEIQVRDUKB-UHFFFAOYSA-M |
Synonym | thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216 |
PubChem CID | 24642 |
ChEBI | CHEBI:37117 |
IUPAC Name | chlorothallium |
SMILES | Cl[Tl] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11318577
|
Thermo Scientific Alfa Aesar
035721.06 |
5 g |
184.92 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11328577
|
Thermo Scientific Alfa Aesar
035721.14 |
25 g |
622.10 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is used as a catalyst in chlorination's and in suntan lamps. The low solubility of TlCl is exploited in chemical synthesis: treatment of metal chloride complexes with TlPF6, gives the corresponding metal hexafluorophosphate derivative. It is used as a tracer in medical imaging.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Chemical Identifiers
7791-12-0 | |
239.83 | |
GBECUEIQVRDUKB-UHFFFAOYSA-M | |
24642 | |
chlorothallium |
ClTl | |
MFCD00011274 | |
thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216 | |
CHEBI:37117 | |
Cl[Tl] |
Specifications
430°C | |
Beads | |
MFCD00011274 | |
14,9260 | |
Slightly soluble in water. Insoluble in alcohol,and acetone. | |
Cl[Tl] | |
239.83 | |
CHEBI:37117 | |
99.999% | |
2.247 | |
7.0 g/mL | |
Thallium(I) chloride, Ultra dry |
7791-12-0 | |
ClTl | |
UN1707 | |
thallium chloride, thallium i chloride, thallous chloride, thallium monochloride, thallium 1+ chloride, tlcl, thallium chloride tlcl, thallium chloride van, rcra waste number u216, rcra waste no. u216 | |
GBECUEIQVRDUKB-UHFFFAOYSA-M | |
chlorothallium | |
24642 | |
239.82 | |
Ampule | |
Air Sensitive | |
(metals basis) |
Safety and Handling
GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c
H300+H330-H373
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.
missing translation for 'einecsNumber' : 232-241-4
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Ampouled under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only