missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) fluoride, 97%, Thermo Scientific Chemicals

198.99 € - 494.46 €

Chemical Identifiers

CAS 7789-27-7
Molecular Formula FTl
Molecular Weight (g/mol) 223.38
MDL Number MFCD00049607
InChI Key CULOEOTWMUCRSJ-UHFFFAOYSA-M
Synonym thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
PubChem CID 62675
SMILES [F-].[Tl+]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11384899
SDS View Documents
Thermo Scientific Alfa Aesar
078111.14
25 g
198.99 €
25g
Please to purchase this item. Need a web account? Register with us today!
11374899
SDS View Documents
Thermo Scientific Alfa Aesar
078111.22
100 g
494.46 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Thallium(I) fluoride is used as chemical reagents, fine chemicals, pharmaceutical research and development.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7789-27-7
223.38
CULOEOTWMUCRSJ-UHFFFAOYSA-M
62675
FTl
MFCD00049607
thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
[F-].[Tl+]

Specifications

655°C
Powder
MFCD00049607
14,9262
Very soluble in water
[F-].[Tl+]
223.38
223.37
8.36 g/mL
7789-27-7
FTl
UN1707
thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
CULOEOTWMUCRSJ-UHFFFAOYSA-M
λ¹-thallanylium fluoride
62675
97%
Thallium (I) fluoride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H373
Fatal if swallowed.
Fatal if inhaled.
May cause damage to organs through prolonged or repeated exposure.

P260-P264b-P270-P271-P284-P301+P310-P304+P340-P310-P314-P330-P501c

H300+H330-H373

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: THALLIUM COMPOUNDS, N.O.S.

missing translation for 'einecsNumber' : 232-154-1

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.