missing translation for 'onlineSavingsMsg'
Learn More
Thallium(I) fluoride, 99% Product Code.: 10155320

Thallium(I) fluoride, 99%

Product Code. 10155320
5 g, Plastic bottle
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 10155320

Brand: Thermo Scientific Acros 312690050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 7789-27-7 | FTl | 223.38 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Boiling Point 655°C
Melting Point 322°C
CAS 7789-27-7
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 98.5% min
Molecular Formula FTl
Linear Formula FTl
MDL Number MFCD00049607
Merck Index 15, 9411
Synonym thallium i fluoride, thallium fluoride, thallium monofluoride, thallium fluoride ic
Solubility Information Solubility in water: 78.6g/100mL (15°C)
InChI Key CULOEOTWMUCRSJ-UHFFFAOYSA-M
SMILES [F-].[Tl+]
IUPAC Name fluorothallium
Molecular Weight (g/mol) 223.38
PubChem CID 62675
Formula Weight 223.37
Percent Purity 99%
Packaging Plastic bottle
Chemical Name or Material Thallium(I) fluoride, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thallium(I) fluoride, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.