missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) hexafluorophosphate(V), 97% min, Thermo Scientific Chemicals

Product Code. 11344577
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11344577

Brand: Thermo Scientific Alfa Aesar 039627.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60969-19-9
Molecular Formula F6PTl
Molecular Weight (g/mol) 349.344
MDL Number MFCD00049807
InChI Key FRZBCOUMLHRKRT-UHFFFAOYSA-N
Synonym thallium hexafluorophosphate, tlpf6, thallium i hexafluorophosphate, thallium 1+ hexafluorophosphate, thallium 1+ hexafluoro-$l^ 5-phosphanuide, $l^ 1-thallanylium hexafluoro-$l^ 5-phosphanuide
PubChem CID 10904204
IUPAC Name thallium(1+);hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.[Tl+]

Specifications

Physical Form Crystalline
Quantity 25 g
UN Number UN1707
Formula Weight 349.34
Percent Purity ≤97%
Chemical Name or Material Thallium(I) hexafluorophosphate(V)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.