missing translation for 'onlineSavingsMsg'
Learn More

Thallium(I) sulfate, 99.5% min (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 7446-18-6
Molecular Formula O4STl2
Molecular Weight (g/mol) 504.816
MDL Number MFCD00011278
InChI Key YTQVHRVITVLIRD-UHFFFAOYSA-L
Synonym thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
PubChem CID 24833
ChEBI CHEBI:81836
IUPAC Name thallium(1+);sulfate
SMILES [O-]S(=O)(=O)[O-].[Tl+].[Tl+]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11333918
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040342.09
10 g N/A
N/A
11303918
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040342.18
50 g N/A
N/A
11323918
View Documents Promotion Details
Thermo Scientific Alfa Aesar
040342.30
250 g N/A
N/A
Description

Description

Thallium(I) sulfate is used as a precursor to thallium(I) sulfide, which exhibits high electrical conductivity when exposed to infrared light. It is further used in photocells. It is also mixed with selenium and arsenic to produce low melting glasses.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7446-18-6
504.816
YTQVHRVITVLIRD-UHFFFAOYSA-L
24833
thallium(1+);sulfate
O4STl2
MFCD00011278
thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
CHEBI:81836
[O-]S(=O)(=O)[O-].[Tl+].[Tl+]

Specifications

(decomposition)
7446-18-6
99.5% min (metals basis)
MFCD00011278
YTQVHRVITVLIRD-UHFFFAOYSA-L
thallium(1+);sulfate
24833
504.8
Thallium(I) sulfate
632°C
Crystalline
O4STl2
thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt
[O-]S(=O)(=O)[O-].[Tl+].[Tl+]
504.816
CHEBI:81836
6.77
Videos
Safety and Handling

Safety and Handling

P260-P264b-P270-P280-P301+P310-P302+P352-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c

H300-H311-H315-H319-H372

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-201-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only