missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thallium(I) sulfate, Honeywell Fluka™
Purum,≥98.0% (RT)
59.43 € - 1100.00 €
Chemical Identifiers
CAS | 7446-18-6 |
---|---|
Molecular Formula | O4STl2 |
Molecular Weight (g/mol) | 504.816 |
MDL Number | MFCD00011278 |
InChI Key | YTQVHRVITVLIRD-UHFFFAOYSA-L |
Synonym | thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt |
PubChem CID | 24833 |
ChEBI | CHEBI:81836 |
IUPAC Name | thallium(1+);sulfate |
SMILES | [O-]S(=O)(=O)[O-].[Tl+].[Tl+] |
Description
- ≥95% for general laboratory use.
Chemical Identifiers
7446-18-6 | |
504.816 | |
YTQVHRVITVLIRD-UHFFFAOYSA-L | |
24833 | |
thallium(1+);sulfate |
O4STl2 | |
MFCD00011278 | |
thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt | |
CHEBI:81836 | |
[O-]S(=O)(=O)[O-].[Tl+].[Tl+] |
Specifications
7446-18-6 | |
Tl2SO4 | |
UN1707 | |
YTQVHRVITVLIRD-UHFFFAOYSA-L | |
thallium(1+);sulfate | |
24833 | |
504.83g/mol | |
6.77g/mL at 25°C |
O4STl2 | |
MFCD00011278 | |
thallous sulfate, thallium sulfate, thallium i sulfate, tharattin, zelio, bonide antzix, th-universal, unii-u9f9qir12t, rcra waste number p115, sulfuric acid, dithallium 1+ salt | |
[O-]S(=O)(=O)[O-].[Tl+].[Tl+] | |
504.816 | |
CHEBI:81836 | |
≥98% (RT) | |
Thallium(I) sulfate |
Safety and Handling
P264-P273-P301 + P310-P314
missing translation for 'rtecsNumber' : XG6800000