missing translation for 'onlineSavingsMsg'
Learn More

Thallium(III) chloride hydrate, Thermo Scientific™

Product Code. 11394248
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13453-33-3
MDL Number MFCD00049600
Synonym Thallic chloride

Specifications

Melting Point 37°C
Physical Form Crystalline Aggregates
Quantity 5 g
UN Number UN1707
Solubility Information It is soluble in water 862g/L
Formula Weight 310.73 (Anhydrous)
Chemical Name or Material Thallium(III) chloride hydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'